Anti-ZC3H8 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB41312-100EA 0 EUR
BIRBORB41312-100
Anti-ZC3H8 Rabbit Polyclonal Antibody
Anticuerpos
Western Blot: 1:500-1:3000; Immunochemistry: 1:100-1:500; Immunofluorescence: 1:100-1:200

Type: Primary
Antigen: ZC3H8 (zinc finger CCCH-type containing 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR