Anti-Calsequestrin 2 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB41202-100EA 0 EUR
BIRBORB41202-100
Anti-Calsequestrin 2 Rabbit Polyclonal Antibody
Anticuerpos
Western Blot: 1:500-1:3000

Type: Primary
Antigen: Calsequestrin 2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR