Anti-DPP8 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB41141-100EA 0 EUR
BIRBORB41141-100
Anti-DPP8 Rabbit Polyclonal Antibody
Anticuerpos
Western Blot: 1:500-1:3000

Type: Primary
Antigen: DPP8 (dipeptidyl-peptidase 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR