Anti-TFEC Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB41108-100EA 0 EUR
BIRBORB41108-100
Anti-TFEC Rabbit Polyclonal Antibody
Anticuerpos
  • Western Blot: 1:500-1:3000

Type: Primary
Antigen: TFEC (transcription factor EC)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR