Anti-ORC2 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB40977-100EA 0 EUR
BIRBORB40977-100
Anti-ORC2 Rabbit Polyclonal Antibody
Anticuerpos
Western Blot: 1:500-1:3000; Immunochemistry: 1:100-1:250

Type: Primary
Antigen: ORC2 (origin recognition complex, subunit 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR