Anti-CAD Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB40832-100EA 0 EUR
BIRBORB40832-100
Anti-CAD Rabbit Polyclonal Antibody
Anticuerpos
Western Blot: 1:500-1:3000

Type: Primary
Antigen: CAD (carbamoyl-phosphate synthetase 2, aspartate transcarbamylase, and dihydroorotase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR