Anti-PPIA Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB27667-100EA 0 EUR
BIRBORB27667-100
Anti-PPIA Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: PPIA (peptidylprolyl isomerase A (cyclophilin A))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR