Anti-FLT1 Rabbit polyclonal antibody

Proveedor: Biorbyt
BIRBORB27611-100EA 0 EUR
BIRBORB27611-100
Anti-FLT1 Rabbit polyclonal antibody
Anticuerpos
Type: Primary
Antigen: FLT1 (fms-related tyrosine kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR