Anti-PIN-Cocktail 2 P504S / p63 / HMW Cytokeratin None Monoclonal Antibody

Proveedor: Biorbyt
BIRBORB27518-5EA 0 EUR
BIRBORB27518-5
Anti-PIN-Cocktail 2 P504S / p63 / HMW Cytokeratin None Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: PIN-Cocktail 2 P504S / p63 / HMW Cytokeratin
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: None
Isotype: IgG+IgG2a+IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR