Anti-ENO2 Mouse monoclonal antibody

Proveedor: Biorbyt
BIRBORB27440-5EA 0 EUR
BIRBORB27440-5
Anti-ENO2 Mouse monoclonal antibody
Anticuerpos
Type: Primary
Antigen: ENO2 (enolase 2 (gamma, neuronal))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR