Anti-Elastin, rat Rabbit polyclonal antibody

Proveedor: Biorbyt
BIRBORB27332-0.5EA 0 EUR
BIRBORB27332-0.5
Anti-Elastin, rat Rabbit polyclonal antibody
Anticuerpos
ImmunofluorescenceA 1:40; Immunochemistry: 1:500; ELISA: > 1:200

Type: Primary
Antigen: Elastin, rat
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR