Anti-ACTA1 Mouse Monoclonal Antibody

Proveedor: Biorbyt
BIRBORB27263-1EA 0 EUR
BIRBORB27263-1
Anti-ACTA1 Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: ACTA1 (actin, alpha 1, skeletal muscle)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR