Anti-KRT5 Mouse Monoclonal Antibody

Proveedor: Biorbyt
BIRBORB27188-1EA 0 EUR
BIRBORB27188-1
Anti-KRT5 Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: KRT5 (keratin 5)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR