Anti-APP Goat Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB26683-0.2EA 0 EUR
BIRBORB26683-0.2
Anti-APP Goat Polyclonal Antibody
Anticuerpos
ELISA: 1:25000; Western Blot: 1:5000

Type: Primary
Antigen: APP (amyloid beta (A4) precursor protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR