Anti-VTN Mouse Monoclonal Antibody

Proveedor: Biorbyt
BIRBORB26303-0.5EA 0 EUR
BIRBORB26303-0.5
Anti-VTN Mouse Monoclonal Antibody
Anticuerpos
ELISA: 1-5 ?g/ml

Type: Primary
Antigen: VTN (vitronectin)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR