Anti-hCG beta Mouse monoclonal antibody

Proveedor: Biorbyt
BIRBORB26075-1EA 0 EUR
BIRBORB26075-1
Anti-hCG beta Mouse monoclonal antibody
Anticuerpos
Type: Primary
Antigen: hCG beta
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR