Anti-A2M Goat Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB25794-10EA 0 EUR
BIRBORB25794-10
Anti-A2M Goat Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: A2M (alpha-2-macroglobulin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR