Anti-Carboxy Methyl Lysine Goat Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB25784-1EA 0 EUR
BIRBORB25784-1
Anti-Carboxy Methyl Lysine Goat Polyclonal Antibody
Anticuerpos
Western Blot: 1:200-1:20000

Type: Primary
Antigen: Carboxy Methyl Lysine
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR