Anti-EGFR Mouse monoclonal antibody

Proveedor: Biorbyt
BIRBORB25502-100EA 0 EUR
BIRBORB25502-100
Anti-EGFR Mouse monoclonal antibody
Anticuerpos
DB: 0.5 ?g/ml; Immunofluorescence: 0.1-1 ?g/ml; ELISA: 0.1 ?g/ml

Type: Primary
Antigen: EGFR (epidermal growth factor receptor)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR