Anti-CU2+-OX LDL Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB25194-100EA 0 EUR
BIRBORB25194-100
Anti-CU2+-OX LDL Rabbit Polyclonal Antibody
Anticuerpos
ELISA: 1:500-1:5000; Immunochemistry: 1:500

Type: Primary
Antigen: CU2+-OX LDL
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR