Anti-UTS2 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB25184-20EA 0 EUR
BIRBORB25184-20
Anti-UTS2 Rabbit Polyclonal Antibody
Anticuerpos
ELISA: 1:3000

Type: Primary
Antigen: UTS2 (urotensin 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR