Anti-TNNT2 Mouse Monoclonal Antibody

Proveedor: Biorbyt
BIRBORB24845-1EA 0 EUR
BIRBORB24845-1
Anti-TNNT2 Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: TNNT2 (troponin T type 2 (cardiac))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR