Anti-APOA1 Mouse Monoclonal Antibody

Proveedor: Biorbyt
BIRBORB24737-1EA 0 EUR
BIRBORB24737-1
Anti-APOA1 Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: APOA1 (apolipoprotein A-I)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR