Anti-Digoxin Mouse monoclonal antibody

Proveedor: Biorbyt
BIRBORB24546-0.2EA 0 EUR
BIRBORB24546-0.2
Anti-Digoxin Mouse monoclonal antibody
Anticuerpos
Type: Primary
Antigen: Digoxin
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR