Anti-POMC Mouse Monoclonal Antibody

Proveedor: Biorbyt
BIRBORB24337-1EA 0 EUR
BIRBORB24337-1
Anti-POMC Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: POMC (proopiomelanocortin)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR