Anti-TSH Goat Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB24170-1EA 0 EUR
BIRBORB24170-1
Anti-TSH Goat Polyclonal Antibody
Anticuerpos
RIA: 1:580000

Type: Primary
Antigen: TSH (Thyroid Stimulating Hormone)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR