Anti-XK Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB24140-100EA 0 EUR
BIRBORB24140-100
Anti-XK Rabbit Polyclonal Antibody
Anticuerpos
Immunochemistry: 1:500

Type: Primary
Antigen: XK (X-linked Kx blood group (McLeod syndrome))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR