Anti-Adenovirus hexon Mouse Monoclonal Antibody

Proveedor: Biorbyt
BIRBORB24093-1EA 0 EUR
BIRBORB24093-1
Anti-Adenovirus hexon Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: Adenovirus hexon
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR