Anti-Fr. tularensis (LPS) Mouse Monoclonal Antibody

Proveedor: Biorbyt
BIRBORB24020-1EA 0 EUR
BIRBORB24020-1
Anti-Fr. tularensis (LPS) Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: Fr. tularensis (LPS)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR