Anti-HSV-1 gG-1 Mouse Monoclonal Antibody
Proveedor: Biorbyt
BIRBORB23914-0.5EA 0 EUR
BIRBORB23914-0.5
Anti-HSV-1 gG-1 Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: HSV-1 gG-1
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now