Anti-Bacteroides Mouse monoclonal antibody

Proveedor: Biorbyt
BIRBORB23338-100EA 0 EUR
BIRBORB23338-100
Anti-Bacteroides Mouse monoclonal antibody
Anticuerpos
Type: Primary
Antigen: Bacteroides
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR