Anti-Anthrax Toxin Recep. Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB23313-100EA 0 EUR
BIRBORB23313-100
Anti-Anthrax Toxin Recep. Rabbit Polyclonal Antibody
Anticuerpos
Western Blot: 1-2 ?g/ml

Type: Primary
Antigen: Anthrax Toxin Recep.
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR