Anti-HBsAg (ad/ay) Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Proveedor: Biorbyt
BIRBORB23232-1EA 0 EUR
BIRBORB23232-1
Anti-HBsAg (ad/ay) Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticuerpos
Type: Primary
Antigen: HBsAg (ad/ay)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now