Anti-N. meningitidis Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB23207-1EA 0 EUR
BIRBORB23207-1
Anti-N. meningitidis Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: N. meningitidis
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR