Anti-HBsAg (ad/ay) Goat Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB23119-1EA 0 EUR
BIRBORB23119-1
Anti-HBsAg (ad/ay) Goat Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: HBsAg (ad/ay)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR