Anti-Cytomegalovirus Rabbit Polyclonal Antibody
Proveedor: Biorbyt
BIRBORB23055-1EA 0 EUR
BIRBORB23055-1
Anti-Cytomegalovirus Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: Cytomegalovirus
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now