Anti-HINT1 Rabbit Polyclonal Antibody
Proveedor: Biorbyt
BIRBORB13467-100EA 0 EUR
BIRBORB13467-100 BIRBORB13467-200
Anti-HINT1 Rabbit Polyclonal Antibody
Anticuerpos
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: HINT1 (histidine triad nucleotide binding protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat