Anti-KISS1R Rabbit Polyclonal Antibody
Proveedor: Biorbyt
BIRBORB13444-100EA 0 EUR
BIRBORB13444-100 BIRBORB13444-200 BIRBORB13444-5 BIRBORB13444-20
Anti-KISS1R Rabbit Polyclonal Antibody
Anticuerpos
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: KISS1R (KISS1 receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat