Anti-FCER2 Rabbit polyclonal antibody

Proveedor: Biorbyt
BIRBORB13308-100EA 312 EUR
BIRBORB13308-100 BIRBORB13308-5 BIRBORB13308-20 BIRBORB13308-50
Anti-FCER2 Rabbit polyclonal antibody
Anticuerpos
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: FCER2 (Fc fragment of IgE, low affinity II, receptor for (CD23))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR