Anti-KIR2DL1 Rabbit Polyclonal Antibody
Proveedor: Biorbyt
BIRBORB13304-200EA 0 EUR
BIRBORB13304-200 BIRBORB13304-100
Anti-KIR2DL1 Rabbit Polyclonal Antibody
Anticuerpos
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: KIR2DL1 (killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human