Anti-AQP7 Rabbit polyclonal antibody

Proveedor: Biorbyt
BIRBORB13253-100EA 312 EUR
BIRBORB13253-100 BIRBORB13253-200
Anti-AQP7 Rabbit polyclonal antibody
Anticuerpos
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: AQP7 (aquaporin 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR