Anti-SHBG Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB13231-200EA 508 EUR
BIRBORB13231-200 BIRBORB13231-100
Anti-SHBG Rabbit Polyclonal Antibody
Anticuerpos
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: SHBG (sex hormone-binding globulin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR