Anti-HSPD1 Mouse Monoclonal Antibody

Proveedor: Biorbyt
BIRBORB13222-100EA 0 EUR
BIRBORB13222-100
Anti-HSPD1 Mouse Monoclonal Antibody
Anticuerpos
Western Blot: 1:500-1000

Type: Primary
Antigen: HSPD1 (heat shock 60kDa protein 1 (chaperonin))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR