Anti-DNTT Mouse Monoclonal Antibody

Proveedor: Biorbyt
BIRBORB13201-100EA 0 EUR
BIRBORB13201-100
Anti-DNTT Mouse Monoclonal Antibody
Anticuerpos
Western Blot: 11000

Type: Primary
Antigen: DNTT (DNA nucleotidylexotransferase)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR