Anti-KIT Mouse Monoclonal Antibody

Proveedor: Biorbyt
BIRBORB13196-100EA 0 EUR
BIRBORB13196-100
Anti-KIT Mouse Monoclonal Antibody
Anticuerpos
Western Blot: 1:1000

Type: Primary
Antigen: KIT (v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR