Anti-KRT17 Mouse Monoclonal Antibody

Proveedor: Biorbyt
BIRBORB13186-100EA 341 EUR
BIRBORB13186-100
Anti-KRT17 Mouse Monoclonal Antibody
Anticuerpos
Western Blot: 1:2000

Type: Primary
Antigen: KRT17 (keratin 17)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR