Anti-APP Rabbit polyclonal antibody

Proveedor: Biorbyt
BIRBORB13182-100EA 341 EUR
BIRBORB13182-100
Anti-APP Rabbit polyclonal antibody
Anticuerpos
  • Western Blot: 1:1000

Type: Primary
Antigen: APP (amyloid beta (A4) precursor protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR