Anti-CD27 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB13179-100EA 0 EUR
BIRBORB13179-100
Anti-CD27 Rabbit Polyclonal Antibody
Anticuerpos
Western Blot: 1:2000-5000

Type: Primary
Antigen: CD27 (CD27 molecule)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR