Anti-MYC Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Proveedor: Biorbyt
BIRBORB13174-100EA 341 EUR
BIRBORB13174-100
Anti-MYC Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticuerpos
Western Blot: 1:1000-2000

Type: Primary
Antigen: MYC (v-myc avian myelocytomatosis viral oncogene homolog)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR