Anti-HA tag Rabbit Polyclonal Antibody

Proveedor: Biorbyt
BIRBORB13168-100EA 0 EUR
BIRBORB13168-100
Anti-HA tag Rabbit Polyclonal Antibody
Anticuerpos
WB1:10000

Type: Primary
Antigen: HA tag (HA tag)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR